siRNA Details
| Gene Symbol: Gm33354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135929.1 | - | 3 | 434 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ052803.1 | - | 3 | 434 | Chr. 1 - 166448875 - 166455488 [-] on Build GRCm38 | |

Gm33354
Genomic Map