siRNA Details
| Gene Symbol: 4933406L23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007295.1 | - | 1 | 653 | ||
| AK016703.1 | - | 1 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406L23Rik| Gene Symbol: 4933406L23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007295.1 | - | 1 | 653 | ||
| AK016703.1 | - | 1 | 647 | |||