siRNA Details
| Gene Symbol: 1700126H18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040695.1 | 3 | 304 | |||
| GenBank mRNA | AK007290.1 | - | 3 | 305 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707392.1 | - | 3 | 305 | Chr. 5 - 66166607 - 66191335 [-] on Build GRCm38 | |

1700126H18Rik
Genomic Map