siRNA Details
| Gene Symbol: 4933411O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016780.1 | - | 1 | 427 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716458.1 | - | 1 | 427 | Chr. 16 - 66892511 - 66893150 [+] on Build GRCm38 | |

4933411O13Rik
Genomic Map