siRNA Details
| Gene Symbol: 1700104L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108033.1 | 3 | 570 | |||
| GenBank mRNA | AK007120.1 | - | 3 | 571 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700104L18Rik| Gene Symbol: 1700104L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108033.1 | 3 | 570 | |||
| GenBank mRNA | AK007120.1 | - | 3 | 571 | ||