siRNA Details
| Gene Symbol: 1700104L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108033.1 | 3 | 364 | |||
| GenBank mRNA | AK007120.1 | - | 3 | 365 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707200.1 | - | 3 | 365 | Chr. 12 - 54233615 - 54242315 [+] on Build GRCm38 | |

1700104L18Rik
Genomic Map