siRNA Details
| Gene Symbol: 1700008B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005752.1 | - | 1 | 87 | ||
| AK007127.1 | - | 1 | 114 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707205.1 | - | 1 | 114 | Chr. 5 - 121835195 - 121835707 [+] on Build GRCm38 | |

1700008B11Rik
Genomic Map