siRNA Details
| Gene Symbol: Gm12108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032705.1 | - | 2 | 287 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB623133.1 | - | 2 | 284 | Chr. 11 - 32097205 - 32101226 [-] on Build GRCm38 | |

Gm12108
Genomic Map