siRNA Details
Gene Symbol: 8430437L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040503.1 | 1 | 384 | |||
GenBank mRNA | AK018472.1 | - | 1 | 384 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720607.1 | - | 384 | Chr. - - [-] on Build |
