siRNA Details
| Gene Symbol: Lppos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037954.1 | 2 | 667 | |||
| GenBank mRNA | AK004252.1 | - | 2 | 455 | ||
| AK039760.1 | - | 473 | ||||
| BC037367.1 | - | 1 | 361 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lppos| Gene Symbol: Lppos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037954.1 | 2 | 667 | |||
| GenBank mRNA | AK004252.1 | - | 2 | 455 | ||
| AK039760.1 | - | 473 | ||||
| BC037367.1 | - | 1 | 361 | |||