siRNA Details
| Gene Symbol: 5730403I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040378.1 | 4 | 1453 | |||
| GenBank mRNA | AK017485.1 | - | 4 | 1457 | ||
| AK161639.1 | - | 4 | 1352 | |||
| BC048919.1 | - | 4 | 1354 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730403I07Rik| Gene Symbol: 5730403I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040378.1 | 4 | 1453 | |||
| GenBank mRNA | AK017485.1 | - | 4 | 1457 | ||
| AK161639.1 | - | 4 | 1352 | |||
| BC048919.1 | - | 4 | 1354 | |||