siRNA Details
| Gene Symbol: 5730403I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040378.1 | 4 | 644 | |||
| GenBank mRNA | AK017485.1 | - | 4 | 646 | ||
| AK161639.1 | - | 4 | 543 | |||
| BC048919.1 | - | 4 | 544 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ715174.1 | - | 4 | 543 | Chr. 9 - 77367445 - 77399315 [-] on Build GRCm38 | |

5730403I07Rik
Genomic Map