siRNA Details
Gene Symbol: 5730403I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040378.1 | 4 | 446 | |||
GenBank mRNA | AK017485.1 | - | 4 | 448 | ||
AK161639.1 | - | 4 | 344 | |||
BC048919.1 | - | 4 | 346 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ715174.1 | - | 4 | 345 | Chr. 9 - 77367445 - 77399315 [-] on Build GRCm38 |
