siRNA Details
| Gene Symbol: 1500026H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005299.1 | - | 1 | 17 | ||
| AK048993.1 | - | 1 | 38 | |||
| AK085365.1 | - | 1 | 15 | |||
| BY260309.1 | - | 1 | 47 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500026H17Rik| Gene Symbol: 1500026H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005299.1 | - | 1 | 17 | ||
| AK048993.1 | - | 1 | 38 | |||
| AK085365.1 | - | 1 | 15 | |||
| BY260309.1 | - | 1 | 47 | |||