siRNA Details
| Gene Symbol: A930038B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050096.1 | - | 1673 | |||
| Gene Symbol: Kin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC058169.1 | - | 4 | 1325 | ||

A930038B10Rik
Genomic Map