siRNA Details
| Gene Symbol: A930038B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050096.1 | - | 1512 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930038B10Rik| Gene Symbol: A930038B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050096.1 | - | 1512 | |||