siRNA Details
| Gene Symbol: 5530401A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038010.1 | 1 | 389 | |||
| GenBank mRNA | AK017430.1 | - | 1 | 157 | ||
| AK029248.1 | - | 1 | 393 | |||
| BB613856.1 | - | 1 | 389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5530401A14Rik| Gene Symbol: 5530401A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038010.1 | 1 | 389 | |||
| GenBank mRNA | AK017430.1 | - | 1 | 157 | ||
| AK029248.1 | - | 1 | 393 | |||
| BB613856.1 | - | 1 | 389 | |||