siRNA Details
| Gene Symbol: 2010308F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045429.1 | 1 | 12 | |||
| GenBank mRNA | AK008545.1 | - | 1 | 14 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708429.1 | - | 1 | 14 | Chr. X - 13234684 - 13247733 [-] on Build GRCm38 | |

2010308F09Rik
Genomic Map