siRNA Details
| Gene Symbol: B930036N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154483.1 | - | 610 | |||
| AK171461.1 | - | 1 | 622 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD560072.1 | - | 1 | 418 | Chr. 1 - 171502427 - 171502960 [-] on Build GRCm38 | |

B930036N10Rik
Genomic Map