siRNA Details
| Gene Symbol: B930036N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047216.1 | - | 1 | 1067 | ||
| AK171461.1 | - | 1 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B930036N10Rik| Gene Symbol: B930036N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047216.1 | - | 1 | 1067 | ||
| AK171461.1 | - | 1 | 119 | |||