siRNA Details
| Gene Symbol: Gm11732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045779.1 | - | 2 | 211 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB643605.1 | - | 2 | 211 | Chr. 11 - 117426178 - 117437591 [+] on Build GRCm38 | |

Gm11732
Genomic Map