siRNA Details
Gene Symbol: 3110052M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014206.1 | - | 4 | 935 | ||
AK161479.1 | - | 4 | 871 | |||
AK162561.1 | - | 1 | 3557 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 3110052M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014206.1 | - | 4 | 935 | ||
AK161479.1 | - | 4 | 871 | |||
AK162561.1 | - | 1 | 3557 |