siRNA Details
| Gene Symbol: Gm3716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045078.1 | 4 | 1204 | |||
| GenBank mRNA | AK044377.1 | - | 4 | 1189 | ||
| BC038278.1 | - | 4 | 1183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3716| Gene Symbol: Gm3716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045078.1 | 4 | 1204 | |||
| GenBank mRNA | AK044377.1 | - | 4 | 1189 | ||
| BC038278.1 | - | 4 | 1183 | |||