siRNA Details
Gene Symbol: Gm3716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045078.1 | 4 | 1400 | |||
GenBank mRNA | AK044377.1 | - | 4 | 1385 | ||
BC038278.1 | - | 4 | 1379 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm3716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045078.1 | 4 | 1400 | |||
GenBank mRNA | AK044377.1 | - | 4 | 1385 | ||
BC038278.1 | - | 4 | 1379 |