siRNA Details
| Gene Symbol: A930012M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020849.1 | - | 1 | 329 | ||
| AK139237.1 | - | 1 | 1650 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930012M21Rik| Gene Symbol: A930012M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020849.1 | - | 1 | 329 | ||
| AK139237.1 | - | 1 | 1650 | |||