siRNA Details
| Gene Symbol: 2900008C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045434.1 | 1, 2 | 65 | |||
| GenBank mRNA | BC027704.1 | - | 1, 2 | 65 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900008C10Rik| Gene Symbol: 2900008C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045434.1 | 1, 2 | 65 | |||
| GenBank mRNA | BC027704.1 | - | 1, 2 | 65 | ||