siRNA Details
| Gene Symbol: 4930471G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015551.1 | - | 1 | 90 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715099.1 | - | 1 | 90 | Chr. 13 - 47667550 - 47668393 [+] on Build GRCm38 | |

4930471G24Rik
Genomic Map