siRNA Details
| Gene Symbol: 4930471M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045980.1 | 3 | 1076 | |||
| GenBank mRNA | AK015554.1 | - | 3 | 1077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930471M09Rik| Gene Symbol: 4930471M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045980.1 | 3 | 1076 | |||
| GenBank mRNA | AK015554.1 | - | 3 | 1077 | ||