siRNA Details
| Gene Symbol: 4931428A05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015556.1 | - | 1 | 366 | ||
| AK016478.1 | - | 1 | 335 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715106.1 | - | 1 | 364 | Chr. 1 - 90758619 - 90759355 [+] on Build GRCm38 | |

4931428A05Rik
Genomic Map