siRNA Details
| Gene Symbol: 1110025M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003925.1 | - | 1 | 392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110025M09Rik| Gene Symbol: 1110025M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003925.1 | - | 1 | 392 | ||