siRNA Details
| Gene Symbol: Gm16876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040110.1 | - | 652 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB205223.2 | - | 4 | 538 | Chr. 12 - 91684780 - 91693590 [+] on Build GRCm38 | |

Gm16876
Genomic Map