siRNA Details
| Gene Symbol: Gm11762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045099.1 | 4 | 1546 | |||
| GenBank mRNA | AK042811.1 | - | 46 | |||
| AK053782.1 | - | 4 | 1549 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11762| Gene Symbol: Gm11762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045099.1 | 4 | 1546 | |||
| GenBank mRNA | AK042811.1 | - | 46 | |||
| AK053782.1 | - | 4 | 1549 | |||