siRNA Details
| Gene Symbol: Gm11762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042811.1 | - | 1531 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB248549.2 | - | 1 | 513 | Chr. 11 - 119569859 - 119570530 [+] on Build GRCm38 | |

Gm11762
Genomic Map