siRNA Details
| Gene Symbol: Gm41640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158473.1 | - | 1 | 68 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ142828.1 | - | 1 | 65 | Chr. 17 - 84163475 - 84164229 [-] on Build GRCm38 | |

Gm41640
Genomic Map