siRNA Details
| Gene Symbol: 1810007D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038136.1 | 3 | 499 | |||
| GenBank mRNA | AK007359.1 | - | 3 | 503 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810007D17Rik| Gene Symbol: 1810007D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038136.1 | 3 | 499 | |||
| GenBank mRNA | AK007359.1 | - | 3 | 503 | ||