siRNA Details
| Gene Symbol: 1810007C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045472.1 | 2 | 411 | |||
| GenBank mRNA | AK007358.1 | - | 2 | 415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810007C17Rik| Gene Symbol: 1810007C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045472.1 | 2 | 411 | |||
| GenBank mRNA | AK007358.1 | - | 2 | 415 | ||