siRNA Details
Gene Symbol: 1810007C17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045472.1 | 1 | 202 | |||
GenBank mRNA | AK007358.1 | - | 1 | 206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810007C17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045472.1 | 1 | 202 | |||
GenBank mRNA | AK007358.1 | - | 1 | 206 |