siRNA Details
Gene Symbol: 1700112J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077218.1 | 2 | 258 | |||
GenBank mRNA | AK018912.1 | - | 2 | 244 | ||
AK018948.1 | - | 2 | 259 | |||
AK028097.1 | - | 2 | 246 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700112J05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077218.1 | 2 | 258 | |||
GenBank mRNA | AK018912.1 | - | 2 | 244 | ||
AK018948.1 | - | 2 | 259 | |||
AK028097.1 | - | 2 | 246 |