siRNA Details
| Gene Symbol: 1700112J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077218.1 | 1 | 53 | |||
| GenBank mRNA | AK018912.1 | - | 1 | 39 | ||
| AK018948.1 | - | 1 | 54 | |||
| AK028097.1 | - | 1 | 41 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700112J05Rik| Gene Symbol: 1700112J05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077218.1 | 1 | 53 | |||
| GenBank mRNA | AK018912.1 | - | 1 | 39 | ||
| AK018948.1 | - | 1 | 54 | |||
| AK028097.1 | - | 1 | 41 | |||