siRNA Details
| Gene Symbol: Gm10832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136598.1 | - | 1, 2 | 209 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720934.1 | - | 1, 2 | 209 | Chr. 16 - 11031956 - 11036420 [+] on Build GRCm38 | |

Gm10832
Genomic Map