siRNA Details
| Gene Symbol: 9530059O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049978.1 | - | 403 | |||
| AK079255.1 | - | 488 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722185.1 | - | 4 | 483 | Chr. 9 - 122572506 - 122673164 [+] on Build GRCm38 | |

9530059O14Rik
Genomic Map