siRNA Details
| Gene Symbol: 9530059O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049978.1 | - | 1032 | |||
| AK079255.1 | - | 1117 | ||||
| AK135014.1 | - | 890 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530059O14Rik| Gene Symbol: 9530059O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049978.1 | - | 1032 | |||
| AK079255.1 | - | 1117 | ||||
| AK135014.1 | - | 890 | ||||