siRNA Details
| Gene Symbol: 9530059O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049978.1 | - | 742 | |||
| AK079255.1 | - | 827 | ||||
| AK135014.1 | - | 600 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719010.1 | - | 4 | 591 | Chr. 9 - 122572506 - 122677505 [+] on Build GRCm38 | |

9530059O14Rik
Genomic Map