siRNA Details
| Gene Symbol: 1700089I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018915.1 | - | 1 | 123 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707091.1 | - | 1 | 123 | Chr. 2 - 154782332 - 154782683 [+] on Build GRCm38 | |

1700089I07Rik
Genomic Map