siRNA Details
| Gene Symbol: 9130015G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035625.1 | - | 402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130015G15Rik| Gene Symbol: 9130015G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035625.1 | - | 402 | |||