siRNA Details
Gene Symbol: 2810032G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012851.1 | - | 293 | |||
AK028030.1 | - | 178 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2810032G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012851.1 | - | 293 | |||
AK028030.1 | - | 178 |