siRNA Details
| Gene Symbol: 2310001H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036632.1 | - | 435 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB629580.1 | - | 3 | 435 | Chr. 6 - 129216078 - 129233974 [-] on Build GRCm38 | |

2310001H17Rik
Genomic Map