siRNA Details
| Gene Symbol: 5430425K12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103550.1 | 5 | 1075 | |||
| GenBank mRNA | AK017339.1 | - | 2 | 796 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717153.1 | - | 2 | 796 | Chr. 13 - 80941059 - 80948576 [+] on Build GRCm38 | |

5430425K12Rik
Genomic Map