siRNA Details
| Gene Symbol: 5430425K12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103550.1 | 5 | 1111 | |||
| GenBank mRNA | AK017339.1 | - | 2 | 832 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430425K12Rik| Gene Symbol: 5430425K12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103550.1 | 5 | 1111 | |||
| GenBank mRNA | AK017339.1 | - | 2 | 832 | ||