siRNA Details
Gene Symbol: 5430421F17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040352.1 | 3 | 408 | |||
GenBank mRNA | AK017331.1 | - | 3 | 408 | ||
AK142918.1 | - | 2 | 1238 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717144.1 | - | 3 | 408 | Chr. 8 - 25304007 - 25306270 [-] on Build GRCm38 |
